5-(3-Bromophenyl)oxazole 98%

Supplier: Ambeed
Danger

A178491-1G A178491-25G A178491-5G
76769-478EA 10 USD
76769-478 76769-480 76769-482
5-(3-Bromophenyl)oxazole 98%
5-(3-Bromophenyl)oxazole

Precaución: for Research Use Only

Formula: C₉H₆BrNO
MDL Number: MFCD05668970
CAS Number: 243455-57-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR