3-(Piperazin-1-yl)propan-1-amine 97%

Supplier: Ambeed
Danger

A177323-100MG A177323-250MG
76769-440EA 54.11 USD
76769-440 76769-442
3-(Piperazin-1-yl)propan-1-amine 97%
3-(Piperazin-1-yl)propan-1-amine

Precaución: for Research Use Only

Formula: C₇H₁₇N₃
MW: 143.23 g/mol
MDL Number: MFCD00129013
CAS Number: 34885-02-4
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR