5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-hydroxy-1-methyl-1H-pyrazole-3-carboxylate

A176067-100MG A176067-10G A176067-1G A176067-250MG A176067-25G A176067-5G
76769-388EA 7 USD
76769-388 76769-390 76769-392 76769-394 76769-396 76769-398
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester 97%
5-Hydroxy-1-methyl-1H-pyrazole-3-carboxylic acid methyl ester

Precaución: for Research Use Only

Formula: C₆H₈N₂O₃
MDL Number: MFCD01851055
CAS Number: 51985-95-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR