5-Bromo-2-(methylamino)pyrimidine 98%

Supplier: Ambeed
Warning

A175987-100MG A175987-10G A175987-1G A175987-250MG A175987-25G A175987-5G
76769-368EA 5 USD
76769-368 76769-370 76769-372 76769-374 76769-376 76769-378
5-Bromo-2-(methylamino)pyrimidine 98%
5-Bromo-2-(methylamino)pyrimidine

Precaución: for Research Use Only

Formula: C₅H₆BrN₃
MDL Number: MFCD07368227
CAS Number: 31402-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR