6-(Diethylamino)nicotinaldehyde 97%

Supplier: Ambeed
Warning

A175869-100MG A175869-250MG A175869-1G
76769-364EA 63 USD
76769-364 76769-366 77600-424
6-(Diethylamino)nicotinaldehyde 97%
6-(Diethylamino)nicotinaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₄N₂O
MW: 178.23 g/mol
MDL Number: MFCD06200894
CAS Number: 578726-67-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR