5-Bromo-2-chloro-4-methoxypyrimidine 97%

Supplier: Ambeed
Warning

A175694-100G A175694-10G A175694-1G A175694-25G A175694-5G
76769-354EA 244.01 USD
76769-354 76769-356 76769-358 76769-360 76769-362
5-Bromo-2-chloro-4-methoxypyrimidine 97%
5-Bromo-2-chloro-4-methoxypyrimidine

Precaución: for Research Use Only

Formula: C₅H₄BrClN₂O
MW: 223.4600 g/mol
MDL Number: MFCD03001373
CAS Number: 57054-92-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR