4-Bromo-2-furoic acid 97%

Supplier: Ambeed
Warning

A175126-250MG A175126-5G A175126-1G
76769-342EA 62.99 USD
76769-342 76769-344 77600-418
4-Bromo-2-furoic acid 97%
4-Bromo-2-furoic acid

Precaución: for Research Use Only

Formula: C₅H₃BrO₃
MDL Number: MFCD00092494
CAS Number: 3439-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR