4-(Hydroxymethyl)pyridine 1-oxide 98+%

Supplier: Ambeed
Warning

A175097-100MG A175097-1G A175097-250MG A175097-5G
76769-332EA 13.8 USD
76769-332 76769-334 76769-336 76769-338
4-(Hydroxymethyl)pyridine 1-oxide 98+%
4-(Hydroxymethyl)pyridine 1-oxide

Precaución: for Research Use Only

Formula: C₆H₇NO₂
MW: 125.12 g/mol
MDL Number: MFCD00006211
CAS Number: 22346-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR