5-(Dimethylamino)-pentanoic acid hydrochloride 95%

Supplier: Ambeed
Warning

Synonyms: 5-(Dimethylamino)pentanoic acid hydrochloride

A175060-100MG A175060-1G A175060-250MG A175060-25G A175060-5G
76769-322EA 18.04 USD
76769-322 76769-324 76769-326 76769-328 76769-330
5-(Dimethylamino)-pentanoic acid hydrochloride 95%
5-(Dimethylamino)-pentanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₅NO₂·ClH
MW: 181.66 g/mol
MDL Number: MFCD00035089
CAS Number: 25726-28-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR