5-Fluoro-3,4-dihydroisoquinolin-1(2H)-one 98%

Supplier: Ambeed
Warning

A173030-250MG A173030-5G
76769-294EA 42 USD
76769-294 76769-296
5-Fluoro-3,4-dihydroisoquinolin-1(2H)-one 98%
5-Fluoro-3,4-dihydroisoquinolin-1(2H)-one

Precaución: for Research Use Only

Formula: C₉H₈FNO
MDL Number: MFCD04114877
CAS Number: 230301-83-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR