5-Amino-2-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 3-Cyano-4-fluoroaniline

A172650-100G A172650-10G A172650-1G A172650-25G A172650-5G
76769-284EA 113 USD
76769-284 76769-286 76769-288 76769-290 76769-292
5-Amino-2-fluorobenzonitrile 98%
5-Amino-2-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₅FN₂
MDL Number: MFCD00277872
CAS Number: 53312-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR