6-Chloroimidazo[1,2-b]pyridazine 97%

Supplier: Ambeed
Warning

A171156-100G A171156-10G A171156-1G A171156-25G A171156-5G
76769-236EA 84 USD
76769-236 76769-238 76769-240 76769-242 76769-244
6-Chloroimidazo[1,2-b]pyridazine 97%
6-Chloroimidazo[1,2-b]pyridazine

Precaución: for Research Use Only

Formula: C₆H₄ClN₃
MDL Number: MFCD07778345
CAS Number: 6775-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR