N(α)-Cbz-D-tryptophan 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Z-D-Trp-OH, N(α)-Benzyloxycarbonyl-D-tryptophan
A170900-100G
A170900-1G
A170900-25G
A170900-5G
76769-224EA
51
USD
76769-224
76769-226
76769-228
76769-230
N(α)-Cbz-D-tryptophan 98%
N(α)-Cbz-D-tryptophan
Precaución: for Research Use Only
Formula:
C₁₉H₁₈N₂O₄ |
MDL Number:
MFCD00037945 CAS Number: 2279-15-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...