2,4-Difluorobenzamide 98%

Supplier: Ambeed
Warning

A170607-100G A170607-10G A170607-1G A170607-25G A170607-5G
76769-214EA 126.25 USD
76769-214 76769-216 76769-218 76769-220 76769-222
2,4-Difluorobenzamide 98%
2,4-Difluorobenzamide

Precaución: for Research Use Only

Formula: C₇H₅F₂NO
MW: 157.12 g/mol
MDL Number: MFCD00015547
CAS Number: 85118-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR