5-(Imidazo[1,2-a]pyridin-6-yl)-6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 98%

Supplier: Ambeed
Warning

Synonyms: Olprinone

A168914-10MG A168914-1MG A168914-250MG A168914-50MG A168914-5MG A168914-100MG
76769-162EA 25 USD
76769-162 76769-164 76769-166 76769-168 76769-170 77598-192
5-(Imidazo[1,2-a]pyridin-6-yl)-6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile 98%
5-(Imidazo[1,2-a]pyridin-6-yl)-6-methyl-2-oxo-1,2-dihydropyridine-3-carbonitrile

Precaución: for Research Use Only

Formula: C₁₄H₁₀N₄O
MDL Number: MFCD25541649
CAS Number: 106730-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR