4,4,5,5-Tetramethyl-2-(2,3,6-trifluorophenyl)-1,3,2-dioxaborolane 95%

Supplier: Ambeed
Warning

A1688681-1G A1688681-250MG A1688681-5G
76769-150EA 217 USD
76769-150 76769-152 76769-154
4,4,5,5-Tetramethyl-2-(2,3,6-trifluorophenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(2,3,6-trifluorophenyl)-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₂H₁₄BF₃O₂
MDL Number: MFCD24039525
CAS Number: 1819398-31-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR