5-Methylpyridine-3-sulfonic acid 97%

Supplier: Ambeed
Warning

A168729-1G A168729-5G
76769-128EA 10 USD
76769-128 76769-130
5-Methylpyridine-3-sulfonic acid 97%
5-Methylpyridine-3-sulfonic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₃S
MDL Number: MFCD00023571
CAS Number: 4808-70-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR