2-Bromo-6-fluorobenzonitrile 98%

Supplier: Ambeed
Warning

A168552-100G A168552-10G A168552-25G A168552-5G
76769-114EA 130.5 USD
76769-114 76769-116 76769-118 76769-120
2-Bromo-6-fluorobenzonitrile 98%
2-Bromo-6-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrFN
MW: 200.01 g/mol
MDL Number: MFCD06797753
CAS Number: 79544-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR