1-((4-Methoxyphenyl)sulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%

Supplier: Ambeed
Warning

A1684323-1G A1684323-250MG A1684323-5G
76769-102EA 118.83 USD
76769-102 76769-104 76769-106
1-((4-Methoxyphenyl)sulfonyl)-1H-pyrrolo[2,3-b]pyridine 95%
1-((4-Methoxyphenyl)sulfonyl)-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₂N₂O₃S
MW: 288.32 g/mol
CAS Number: 2485020-27-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR