2,4-Bis(benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine 95%

Supplier: Ambeed
Warning

A1684313-100MG A1684313-1G A1684313-250MG
76769-096EA 33 USD
76769-096 76769-098 76769-100
2,4-Bis(benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine 95%
2,4-Bis(benzyloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidine

Precaución: for Research Use Only

Formula: C₂₄H₂₇BN₂O₄
MW: 418.29 g/mol
MDL Number: MFCD28399193
CAS Number: 2640480-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR