1-(5-Bromoindolin-1-yl)-2-chloroethan-1-one 95%

Supplier: Ambeed
Danger

A1683267-1G A1683267-250MG A1683267-5G
76769-090EA 200.52 USD
76769-090 76769-092 76769-094
1-(5-Bromoindolin-1-yl)-2-chloroethan-1-one 95%
1-(5-Bromoindolin-1-yl)-2-chloroethan-1-one

Precaución: for Research Use Only

Formula: C₁₀H₉BrClNO
MW: 274.54 g/mol
MDL Number: MFCD06778214
CAS Number: 87866-10-2
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR