1-(5-Bromoindolin-1-yl)-2-morpholinoethan-1-one 95%

Supplier: Ambeed
Warning

A1683263-100MG A1683263-1G A1683263-250MG
76769-084EA 46.68 USD
76769-084 76769-086 76769-088
1-(5-Bromoindolin-1-yl)-2-morpholinoethan-1-one 95%
1-(5-Bromoindolin-1-yl)-2-morpholinoethan-1-one

Precaución: for Research Use Only

Formula: C₁₄H₁₇BrN₂O₂
MW: 325.2 g/mol
MDL Number: MFCD09877335
CAS Number: 948355-77-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR