3-(4-Methoxyphenyl)oxetane-3-sulfonyl fluoride 95%

Supplier: Ambeed
Danger

A1682795-100MG A1682795-250MG
76769-074EA 137.92 USD
76769-074 76769-076
3-(4-Methoxyphenyl)oxetane-3-sulfonyl fluoride 95%
3-(4-Methoxyphenyl)oxetane-3-sulfonyl fluoride

Precaución: for Research Use Only

Formula: C₁₀H₁₁FO₄S
MW: 246.26 g/mol
CAS Number: 2761830-88-8
UN: 3261
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR