2-(6-Isopropoxynaphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Warning
|
A1681918-100MG
A1681918-1G
A1681918-250MG
A1681918-5G
76769-044EA
47.75
USD
76769-044
76769-046
76769-048
76769-050
2-(6-Isopropoxynaphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(6-Isopropoxynaphthalen-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₉H₂₅BO₃ MW: 312.21 g/mol |
MDL Number:
MFCD30288517 CAS Number: 2246680-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...