9-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acridine 95%

Supplier: Ambeed
Warning

A1677721-100MG A1677721-250MG A1677721-1G
76769-002EA 118 USD
76769-002 76769-004 77876-663
9-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acridine 95%
9-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)acridine

Precaución: for Research Use Only

Formula: C₁₉H₂₀BNO₂
MDL Number: MFCD11520505
CAS Number: 890042-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR