Direct blue 1 50% (dye content)

Supplier: Ambeed
Warning

Synonyms: Chicago Sky Blue 6B, Pontamine Sky Blue 6B, 6,6'-((3,3'-Dimethoxy-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(4-amino-5-hydroxynaphthalene-1,3-disulphonic acid), tetrasodium salt

A167714-1MG A167714-5G
76768-978EA 5.31 USD
76768-978 76768-980
Direct blue 1 50% (dye content)
Direct blue 1

Precaución: for Research Use Only

Formula: C₃₄H₂₈N₆O₁₆S₄
MW: 992.79 g/mol
MDL Number: MFCD00004020
CAS Number: 2610-05-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR