6-Methyl-1H-indazole 96%

Supplier: Ambeed
Warning

A167284-1G A167284-250MG A167284-5G
76768-922EA 16.98 USD
76768-922 76768-924 76768-926
6-Methyl-1H-indazole 96%
6-Methyl-1H-indazole

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.1625 g/mol
MDL Number: MFCD07781625
CAS Number: 698-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR