5-Methoxyindole-2,7-diboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 5-Methoxy-2,7-bis(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-indole

A1669848-1G A1669848-250MG A1669848-5G
76768-896EA 209 USD
76768-896 76768-898 76768-900
5-Methoxyindole-2,7-diboronic acid pinacol ester 98%
5-Methoxyindole-2,7-diboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₂₁H₃₁B₂NO₅
MDL Number: MFCD15474883
CAS Number: 1256360-22-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR