4,4,5,5-Tetramethyl-2-(3-(neopentyloxy)phenyl)-1,3,2-dioxaborolane 95%
Supplier: Ambeed
|
Warning
|
A1669656-1G
A1669656-250MG
A1669656-5G
76768-890EA
79.57
USD
76768-890
76768-892
76768-894
4,4,5,5-Tetramethyl-2-(3-(neopentyloxy)phenyl)-1,3,2-dioxaborolane 95%
4,4,5,5-Tetramethyl-2-(3-(neopentyloxy)phenyl)-1,3,2-dioxaborolane
Precaución: for Research Use Only
Formula:
C₁₇H₂₇BO₃ MW: 290.20 g/mol |
CAS Number:
2246560-92-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...