5-Bromocytidine 95%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-5-bromo-1-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidin-2(1H)-one

A166879-100MG A166879-1G A166879-1MG A166879-5G
76768-870EA 8.49 USD
76768-870 76768-872 76768-874 76768-876
5-Bromocytidine 95%
5-Bromocytidine

Precaución: for Research Use Only

Formula: C₉H₁₂BrN₃O₅
MW: 322.1127 g/mol
MDL Number: MFCD00057461
CAS Number: 3066-86-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR