(S)-2-(4-(tert-Butyl)-4,5-dihydrooxazol-2-yl)phenol 95%

Supplier: Ambeed
Warning

A1665848-100MG A1665848-250MG
76768-850EA 197.33 USD
76768-850 77625-636
(S)-2-(4-(tert-Butyl)-4,5-dihydrooxazol-2-yl)phenol 95%
(S)-2-(4-(tert-Butyl)-4,5-dihydrooxazol-2-yl)phenol

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₂
MW: 219.28 g/mol
CAS Number: 135948-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR