5-Methylbenzo[d]oxazole-2(3H)-thione 98%

Supplier: Ambeed
Warning

A165857-1G A165857-25G A165857-5G
76768-824EA 16.98 USD
76768-824 76768-826 76768-828
5-Methylbenzo[d]oxazole-2(3H)-thione 98%
5-Methylbenzo[d]oxazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₈H₇NOS
MW: 165.21 g/mol
MDL Number: MFCD00052492
CAS Number: 22876-22-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR