5-Allyl-4,6-dichloropyrimidine 95%

Supplier: Ambeed
Warning

A165773-1G A165773-250MG A165773-5G
76768-806EA 13.8 USD
76768-806 76768-808 76768-810 76768-812
5-Allyl-4,6-dichloropyrimidine 95%
5-Allyl-4,6-dichloropyrimidine

Precaución: for Research Use Only

Formula: C₇H₆Cl₂N₂
MDL Number: MFCD11053501
CAS Number: 16019-31-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR