Guanosine-5'-monophosphate disodium salt 98% +

Supplier: Ambeed
Warning

Synonyms: Disodium 5'-guanylate, 5'-Guanosine monophosphate disodium salt, GMP disodium salt, 5'-GMP 2Na, 5'-Guanylic acid disodium salt, Guanosine-5'-Monophosphate

A165471-1000G A165471-100G A165471-25G A165471-500G
76768-790EA 265 USD
76768-790 76768-792 76768-794 76768-796
Guanosine-5'-monophosphate disodium salt 98% +
Guanosine-5'-monophosphate disodium salt

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₅Na₂O₈P
MW: 407.187 g/mol
MDL Number: MFCD00151238
CAS Number: 5550-12-9

Order Now

Specification Test Results

Loss on drying ≤ 12%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR