2-Amino-6-bromoquinazoline 98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromoquinazolin-2-amine

A164100-250MG A164100-1G A164100-5G
76768-738EA 14 USD
76768-738 77625-614 77877-080
2-Amino-6-bromoquinazoline 98%
2-Amino-6-bromoquinazoline

Precaución: for Research Use Only

Formula: C₈H₆BrN₃
MDL Number: MFCD04114089
CAS Number: 190273-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR