2-(Trifluoromethoxy)benzenesulfonamide 98%

Supplier: Ambeed
Warning

A163822-1G A163822-25G A163822-5G
76768-728EA 11.67 USD
76768-728 76768-730 76768-732 77877-079
2-(Trifluoromethoxy)benzenesulfonamide 98%
2-(Trifluoromethoxy)benzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₆F₃NO₃S
MDL Number: MFCD01320751
CAS Number: 37526-59-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR