2-Fluoro-5-nitrotoluene 98%

Supplier: Ambeed
Warning

Synonyms: 1-Fluoro-2-methyl-4-nitrobenzene

A163600-1000G A163600-100G A163600-10G A163600-25G A163600-500G
76768-710EA 118 USD
76768-710 76768-712 76768-714 76768-716 76768-718
2-Fluoro-5-nitrotoluene 98%
2-Fluoro-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MDL Number: MFCD00007284
CAS Number: 455-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR