2-Amino-5-cyano-3-methylpyridine 98%

Supplier: Ambeed
Warning

A163368-1G A163368-250MG
76768-706EA 45 USD
76768-706 76768-708
2-Amino-5-cyano-3-methylpyridine 98%
2-Amino-5-cyano-3-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MDL Number: MFCD06637357
CAS Number: 183428-91-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR