Dibenzo[b,d]furan-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A163276-1G A163276-250MG A163276-5G
76768-698EA 63 USD
76768-698 76768-700 76768-702
Dibenzo[b,d]furan-4-carboxylic acid 95%
Dibenzo[b,d]furan-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₈O₃
MDL Number: MFCD00092335
CAS Number: 2786-05-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR