Ethyl 5-hydroxynicotinate 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Ethoxycarbonyl)-5-hydroxypyridine, Ethyl 5-hydroxypyridine-3-carboxylate

A163219-1G A163219-250MG A163219-5G
76768-692EA 21.22 USD
76768-692 76768-694 76768-696
Ethyl 5-hydroxynicotinate 97%
Ethyl 5-hydroxynicotinate

Precaución: for Research Use Only

Formula: C₈H₉NO₃
MW: 167.162 g/mol
MDL Number: MFCD00510027
CAS Number: 59288-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR