6-Amino-3-cyano-2-methylpyridine 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-2-methylnicotinonitrile, 6-Amino-3-cyano-2-methylpyridine, 6-Amino-2-methylpyridine-3-carbonitrile

A163175-1G A163175-250MG A163175-5G
76768-686EA 51.99 USD
76768-686 76768-688 76768-690
6-Amino-3-cyano-2-methylpyridine 97%
6-Amino-3-cyano-2-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MW: 133.15 g/mol
MDL Number: MFCD05865116
CAS Number: 183428-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR