5-Bromo-2-methoxyphenylacetonitrile 95%

Supplier: Ambeed
Warning

Synonyms: 2-(5-Bromo-2-methoxyphenyl)acetonitrile

A163078-100MG A163078-1G A163078-250MG
76768-680EA 8.49 USD
76768-680 76768-682 76768-684
5-Bromo-2-methoxyphenylacetonitrile 95%
5-Bromo-2-methoxyphenylacetonitrile

Precaución: for Research Use Only

Formula: C₉H₈BrNO
MW: 226.08 g/mol
MDL Number: MFCD00016392
CAS Number: 7062-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR