3-Bromo-α,α,α-trifluoro-5-nitrotoluene 95%

Supplier: Ambeed

A163067-1000G A163067-100G A163067-10G A163067-25G A163067-500G A163067-5G
76768-668EA 197 USD
76768-668 76768-670 76768-672 76768-674 76768-676 76768-678
3-Bromo-α,α,α-trifluoro-5-nitrotoluene 95%
3-Bromo-α,α,α-trifluoro-5-nitrotoluene

Precaución: for Research Use Only

Formula: C₇H₃BrF₃NO₂
MDL Number: MFCD03788719
CAS Number: 630125-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR