5-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A162913-1G A162913-5G
76768-648EA 29 USD
76768-648 76768-650
5-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid 95%
5-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆Cl₂N₂O₂
MDL Number: MFCD03030206
CAS Number: 276684-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR