2-Bromo-5-fluoro-4-methylbenzaldehyde 97%

Supplier: Ambeed
Warning

A162806-100MG A162806-1G A162806-250MG A162806-5G
76768-640EA 8 USD
76768-640 76768-642 76768-644 76768-646
2-Bromo-5-fluoro-4-methylbenzaldehyde 97%
2-Bromo-5-fluoro-4-methylbenzaldehyde

Precaución: for Research Use Only

Formula: C₈H₆BrFO
MDL Number: MFCD07782047
CAS Number: 916792-21-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR