2-Fluoro-4-nitro-1-(trifluoromethyl)benzene 98%

Supplier: Ambeed
Warning

A162370-1G A162370-250MG A162370-5G
76768-606EA 37 USD
76768-606 76768-608 77625-580
2-Fluoro-4-nitro-1-(trifluoromethyl)benzene 98%
2-Fluoro-4-nitro-1-(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₇H₃F₄NO₂
MDL Number: MFCD07782060
CAS Number: 69411-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR