4-Bromo-2-fluoro-5-methylaniline 98%

Supplier: Ambeed
Warning

A162025-100G A162025-10G A162025-1G A162025-25G A162025-5G
76768-594EA 159 USD
76768-594 76768-596 76768-598 76768-600 76768-602 76768-604
4-Bromo-2-fluoro-5-methylaniline 98%
4-Bromo-2-fluoro-5-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MDL Number: MFCD04973744
CAS Number: 418762-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR