2,6-Dichloro-4-(trifluoromethyl)aniline 98%

Supplier: Ambeed
Danger

A161901-100G A161901-10G A161901-500G A161901-5G
76768-586EA 40 USD
76768-586 76768-588 76768-590 76768-592 77877-069
2,6-Dichloro-4-(trifluoromethyl)aniline 98%
2,6-Dichloro-4-(trifluoromethyl)aniline

Precaución: for Research Use Only

Formula: C₇H₄Cl₂F₃N
MDL Number: MFCD00052918
CAS Number: 24279-39-8
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR