4-Fluorophthalonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluorobenzene-1,2-dicarbonitrile, 4-Fluoro-1,2-benzenedicarbonitrile, 4-Fluoro-1,2-dicyanobenzene

A160877-1G A160877-250MG A160877-25G
76768-552EA 22.28 USD
76768-552 76768-554 76768-556
4-Fluorophthalonitrile 97%
4-Fluorophthalonitrile

Precaución: for Research Use Only

Formula: C₈H₃FN₂
MW: 146.12 g/mol
MDL Number: MFCD00210430
CAS Number: 65610-14-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR