2,4-Dichloro-5-nitrobenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 2,4-Dichloro-5-nitrobenzotrifluoride, 1,3-Dichloro-5-nitro-4-(trifluoromethyl)benzene

A160239-100G A160239-10G A160239-25G A160239-5G
76768-526EA 75.33 USD
76768-526 76768-528 76768-530 76768-532
2,4-Dichloro-5-nitrobenzotrifluoride 98%
2,4-Dichloro-5-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₂Cl₂F₃NO₂
MW: 260 g/mol
MDL Number: MFCD00007073
CAS Number: 400-70-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR